missing translation for 'onlineSavingsMsg'
Learn More

Tri-p-tolyl Phosphate 98%, ACROS Organics™

Product Code. 10371591 Shop All Thermo Scientific Chemicals Products
Click to view available options
:
25g; Glass bottle
This item is not returnable. View return policy

Product Code. 10371591

Brand: Thermo Scientific Acros 422300250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 78-32-0
Melting Point 74°C to 78°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C21H21O4P
MDL Number MFCD00041908
Quantity 25 g
Synonym tri-p-cresyl phosphate, tri-p-tolyl phosphate, tris 4-methylphenyl phosphate, tpcp, phosphoric acid, tris 4-methylphenyl ester, phosphoric acid, tri-p-tolyl ester, phosphoric acid, tri 4-tolyl ester, phosphoric acid, tris methylphenyl ester, p-tolyl phosphate c7h7o 3po, tris-p-tolyl phosphate
InChI Key BOSMZFBHAYFUBJ-UHFFFAOYSA-N
SMILES CC1=CC=C(OP(=O)(OC2=CC=C(C)C=C2)OC2=CC=C(C)C=C2)C=C1
IUPAC Name tris(4-methylphenyl) phosphate
Molecular Weight (g/mol) 368.37
PubChem CID 6529
Formula Weight 368.36
Percent Purity ≥97.5%
Physical Form Powder
Chemical Name or Material Tri-p-tolyl phosphate, 98%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.