missing translation for 'onlineSavingsMsg'
Learn More

Tri(o-tolyl)phosphine, 98+%

Product Code. 11440013
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11440013 1 g 1g
11450013 5 g 5g
11460013 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11440013 Supplier Thermo Scientific Alfa Aesar Supplier No. A12093.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6163-58-2 | C21H21P | 304.373 g/mol

Tri(o-tolyl)phosphine is used in a ruthenium-catalyzed direct amination of alcohols. It is also used in Suzuki reaction. Further, it is used in the preparation of tri-ortho-phosphinselenide by reacting with selenium as a reagent. In addition to this, it acts as a ligand in coordination chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tri(o-tolyl)phosphine is used in a ruthenium-catalyzed direct amination of alcohols. It is also used in Suzuki reaction. Further, it is used in the preparation of tri-ortho-phosphinselenide by reacting with selenium as a reagent. In addition to this, it acts as a ligand in coordination chemistry.

Solubility
Soluble in alcohol. Slightly soluble in cold water. Insoluble in water.

Notes
Light sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6163-58-2
Molecular Formula C21H21P
Molecular Weight (g/mol) 304.373
MDL Number MFCD00008514
InChI Key COIOYMYWGDAQPM-UHFFFAOYSA-N
Synonym tri-o-tolylphosphine, tri o-tolyl phosphine, tris 2-methylphenyl phosphine, tris 2-methylphenyl phosphane, phosphine, tris 2-methylphenyl, tris o-tolyl phosphine, tri-ortho-toylphosphine, tri-ortho-tolylphosphine, phosphine, tri-o-tolyl, tri 2-methylphenyl phosphine
PubChem CID 80271
IUPAC Name tris(2-methylphenyl)phosphane
SMILES CC1=CC=CC=C1P(C2=CC=CC=C2C)C3=CC=CC=C3C

Specifications

Melting Point 122°C to 127°C
Quantity 1 g
Beilstein 661212
Sensitivity Air Sensitive
Solubility Information Soluble in alcohol. Slightly soluble in cold water. Insoluble in water.
Formula Weight 304.37
Percent Purity ≥98%
Chemical Name or Material Tri (o-tolyl)phosphine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.