missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -25°C |
| Color | Colorless to Yellow |
| Density | 1.2000g/mL |
| Boiling Point | 410°C |
| Flash Point | 225°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 95% min. (GC) |
| Packaging | Glass bottle |
| Refractive Index | 1.5580 to 1.561 |
| Quantity | 500 g |
| Show More |
Safety and Handling
Product Identifier
- Tri-o-tolyl phosphate
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 2
- Specific target organ toxicity after single exposure Category 1
- H411-Toxic to aquatic life with long lasting effects.
- H370-Causes damage to organs.
- P273-Avoid release to the environment.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?