missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-propyltin chloride

Product Code. 11394809
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11394809 1 g 1g
11384809 5 g 5g
11374809 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11394809 Supplier Thermo Scientific Alfa Aesar Supplier No. 071122.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2279-76-7 | C9H21ClSn | 283.427 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2279-76-7
Molecular Formula C9H21ClSn
Molecular Weight (g/mol) 283.427
MDL Number MFCD00013610
InChI Key FVFLIQMADUVDSP-UHFFFAOYSA-M
Synonym tri-n-propyltin chloride, tripropyltin chloride, stannane, chlorotripropyl, tripropylstannium chloride, tin, tripropyl-, chloride, unii-7iq49g05u7, chloro tripropyl stannane, tri-n-propyl tin chloride, chlorotripropyl stannane, tri-n-propyltinchloride
PubChem CID 16784
IUPAC Name chloro(tripropyl)stannane
SMILES CCC[Sn](CCC)(CCC)Cl

Specifications

Melting Point -23.5°C
Density 1.2678 g/mL at 28°C
Boiling Point 123°C (13 mmHg)
Refractive Index 1.491
Quantity 1 g
UN Number UN2788
Sensitivity Moisture sensitive
Formula Weight 283.41
Physical Form Liquid
Chemical Name or Material Tri-n-propyltin chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.