missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-propylamine, 98%

Product Code. 11404023
Change view
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11404023 250 mL 250mL
11433900 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11404023 Supplier Thermo Scientific Alfa Aesar Supplier No. A10285.AK
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 102-69-2 | C9H21N | 143.27 g/mol

Tri-n-propylamine is used as an chemical intermediate, organic solvent. It is used in the preparation of chemical compounds such as dyes, drugs, catalyst & other uses. It is used together with ruthenium in the electrochemiluminescence.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tri-n-propylamine is used as an chemical intermediate, organic solvent. It is used in the preparation of chemical compounds such as dyes, drugs, catalyst & other uses. It is used together with ruthenium in the electrochemiluminescence.

Solubility
Soluble in water (2.6 g/L) at 20°C, ether and ethanol.

Notes
Hygroscopic. Incompatible with oxidizing agents, acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-69-2
Molecular Formula C9H21N
Molecular Weight (g/mol) 143.27
MDL Number MFCD00009363
InChI Key YFTHZRPMJXBUME-UHFFFAOYSA-N
Synonym tripropylamine, tri-n-propylamine, n,n-dipropyl-1-propanamine, 1-propanamine, n,n-dipropyl, propyldi-n-propylamine, tripropylammonium, tripropyl amine, npr3, n,n-dipropyl-1-propylamine, tripropylamin
PubChem CID 7616
ChEBI CHEBI:38880
IUPAC Name N,N-dipropylpropan-1-amine
SMILES CCCN(CCC)CCC

Specifications

Melting Point -93°C
Density 0.755
Boiling Point 156°C
Flash Point 34°C (93°F)
Odor Amine-like
Linear Formula (CH3CH2CH2)3N
Refractive Index 1.416
Quantity 250 mL
UN Number UN2260
Sensitivity Hygroscopic
Solubility Information Soluble in water (2.6g/L) at 20°C,ether and ethanol.
Formula Weight 143.27
Percent Purity 98%
Chemical Name or Material Tri-n-propylamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tri-n-propylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Tripropylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.