missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-propylamine, 98%

Product Code. 11433900
Change view
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11433900 1 L 1 L
11404023 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11433900 Supplier Thermo Scientific Alfa Aesar Supplier No. A10285.K2
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 102-69-2 | C9H21N | 143.27 g/mol

Tri-n-propylamine is used as an chemical intermediate, organic solvent. It is used in the preparation of chemical compounds such as dyes, drugs, catalyst & other uses. It is used together with ruthenium in the electrochemiluminescence.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tri-n-propylamine is used as an chemical intermediate, organic solvent. It is used in the preparation of chemical compounds such as dyes, drugs, catalyst & other uses. It is used together with ruthenium in the electrochemiluminescence.

Solubility
Soluble in water (2.6 g/L) at 20°C, ether and ethanol.

Notes
Hygroscopic. Incompatible with oxidizing agents, acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-69-2
Molecular Formula C9H21N
Molecular Weight (g/mol) 143.27
MDL Number MFCD00009363
InChI Key YFTHZRPMJXBUME-UHFFFAOYSA-N
Synonym tripropylamine, tri-n-propylamine, n,n-dipropyl-1-propanamine, 1-propanamine, n,n-dipropyl, propyldi-n-propylamine, tripropylammonium, tripropyl amine, npr3, n,n-dipropyl-1-propylamine, tripropylamin
PubChem CID 7616
ChEBI CHEBI:38880
IUPAC Name N,N-dipropylpropan-1-amine
SMILES CCCN(CCC)CCC

Specifications

Melting Point -93°C
Density 0.755
Boiling Point 156°C
Flash Point 34°C (93°F)
Odor Amine-like
Linear Formula (CH3CH2CH2)3N
Refractive Index 1.416
Quantity 1 L
UN Number UN2260
Sensitivity Hygroscopic
Solubility Information Soluble in water (2.6g/L) at 20°C,ether and ethanol.
Formula Weight 143.27
Percent Purity 98%
Chemical Name or Material Tri-n-propylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.