missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-propyl borate, 98%

Product Code. p-7049808
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
This item is not returnable. View return policy

Product Code. 11352588

Brand: Thermo Scientific Alfa Aesar 022974.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 688-71-1 | C9H21BO3 | 188.074 g/mol

Tri-n-propyl borate is used as solvents or catalysts for the production of resins, waxes, paints and varnishes, antioxidants, flame retardants. It is also used for petroleum products, tackifiers for silicone elastomers, stabilizers and binders for rubbers or plastics and as cross-linking agents in polymerization processes. They are are used in welding fluxes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 688-71-1
Molecular Formula C9H21BO3
Molecular Weight (g/mol) 188.074
MDL Number MFCD00009366
InChI Key LTEHWCSSIHAVOQ-UHFFFAOYSA-N
Synonym tri-n-propyl borate, tripropoxyborane, boron propoxide, propyl orthoborate, tris propoxy borane, boric acid h3bo3 , tripropyl ester, tripropyl orthoborate, boric acid, tripropyl ester, propyl borate, pro 3b, boric acid tri-n-propyl ester
PubChem CID 12711
IUPAC Name tripropyl borate
SMILES B(OCCC)(OCCC)OCCC

Specifications

Color Colorless
Density 0.861g/mL
Boiling Point 175°C to 177°C
Flash Point 32° (90°F)
Linear Formula (C3H7O)3B
Refractive Index 1.3950
Quantity 50 g
UN Number UN3272
Sensitivity Moisture Sensitive
Solubility Information Soluble in organic solvents; Decomposes in water
Formula Weight 188.08
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Tri-n-propyl borate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.