missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tri-n-propyl borate, 98%, Thermo Scientific Chemicals
85.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
50 g
250 g
1 kg
Description
Tri-n-propyl borate is used as solvents or catalysts for the production of resins, waxes, paints and varnishes, antioxidants, flame retardants. It is also used for petroleum products, tackifiers for silicone elastomers, stabilizers and binders for rubbers or plastics and as cross-linking agents in polymerization processes. They are are used in welding fluxes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Colorless |
Density | 0.861 g/mL |
Boiling Point | 175°C to 177°C |
Flash Point | 32°C (90°F) |
Linear Formula | (C3H7O)3B |
Refractive Index | 1.395 |
Quantity | 250 g |
UN Number | UN3272 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in organic solvents; Decomposes in water |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'