missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-octylphosphine oxide, 98%

Product Code. 11448913
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11448913 100 g 100g
11438913 25 g 25g
11458913 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11448913 Supplier Thermo Scientific Alfa Aesar Supplier No. A11062.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 78-50-2 | C24H51OP | 386.645 g/mol

Tri-n-octylphosphine oxide is a solvent used in the extraction of metals, especially uranium, zirconium and hafnium. It is also used as an auxiliary reagent in the solvent extraction of metals. It is also used to extract hydrogen bonding organic compounds. Further, it is useful as a capping ligand for the production of quantum dots, such as those consisting of cadmium selenide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tri-n-octylphosphine oxide is a solvent used in the extraction of metals, especially uranium, zirconium and hafnium. It is also used as an auxiliary reagent in the solvent extraction of metals. It is also used to extract hydrogen bonding organic compounds. Further, it is useful as a capping ligand for the production of quantum dots, such as those consisting of cadmium selenide.

Solubility
Soluble in chloroform, toluene and hexane. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-50-2
Molecular Formula C24H51OP
Molecular Weight (g/mol) 386.645
MDL Number MFCD00002083
InChI Key ZMBHCYHQLYEYDV-UHFFFAOYSA-N
Synonym trioctylphosphine oxide, tri-n-octylphosphine oxide, topo, phosphine oxide, trioctyl, cyanex 921, hostarex px 324, trioctyl phosphine oxide, trioctylphosphane oxide, trioctylphosphino-1-one, acmc-209peg
PubChem CID 65577
IUPAC Name 1-dioctylphosphoryloctane
SMILES CCCCCCCCP(=O)(CCCCCCCC)CCCCCCCC

Specifications

Melting Point 52°C to 57°C
Density 0.88
Boiling Point 212°C to 213°C (0.3 mmHg)
Flash Point 252°C (486°F)
Odor Odorless
Quantity 100 g
Beilstein 1796648
Solubility Information Soluble in chloroform,toluene and hexane. Insoluble in water.
Formula Weight 386.65
Percent Purity 98%
Chemical Name or Material Tri-n-octylphosphine oxide
Show More Show Less
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.