missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-hexylamine, 96%

Product Code. 11480138
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11480138 50 g 50g
11490138 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11480138 Supplier Thermo Scientific Alfa Aesar Supplier No. B23716.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 102-86-3 | C18H39N | 269.517 g/mol

Tri-n-hexylamine is used as an extractant during isolation and purification of succinic acid from an escherichia coli fermentation broth in pharmaceutical industry. It is also used as an internal standard in screening of human hair specimens for designer drugs by ion mobility spectrometry. Further, it is involved in the synthesis of zinc oxide nanorods. In addition to this, it is used in the preparation of infrared luminescence nanohybrid films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tri-n-hexylamine is used as an extractant during isolation and purification of succinic acid from an escherichia coli fermentation broth in pharmaceutical industry. It is also used as an internal standard in screening of human hair specimens for designer drugs by ion mobility spectrometry. Further, it is involved in the synthesis of zinc oxide nanorods. In addition to this, it is used in the preparation of infrared luminescence nanohybrid films.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents, acids, acid chloridres, acid anhydrides and chloroformates.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-86-3
Molecular Formula C18H39N
Molecular Weight (g/mol) 269.517
MDL Number MFCD00009523
InChI Key DIAIBWNEUYXDNL-UHFFFAOYSA-N
Synonym tri-n-hexylamine, trihexylamine, 1-hexanamine, n,n-dihexyl, c6-c12 trialkylamine, amines, tri-c6-12-alkyl, amine,trihexyl, 1-hexanamine,n-dihexyl, 1-hexanamine,n,n-dihexyl
PubChem CID 66022
IUPAC Name N,N-dihexylhexan-1-amine
SMILES CCCCCCN(CCCCCC)CCCCCC

Specifications

Melting Point <-75°C
Density 0.798
Boiling Point 263°C to 265°C
Flash Point 144°C (291°F)
Refractive Index 1.4415
Quantity 50 g
UN Number UN3082
Beilstein 1755828
Solubility Information Slightly miscible with water.
Formula Weight 269.52
Percent Purity 97%
Chemical Name or Material Tri-n-hexylamine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tri-n-hexylamine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: N,N-dihexyl-1-hexanamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.