missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-hexyl phosphate, 90+%

Product Code. 11357218
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11357218 5 g 5g
11440490 1 g 1g
11367218 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11357218 Supplier Thermo Scientific Alfa Aesar Supplier No. 016801.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2528-39-4 | C18H39O4P | 350.48 g/mol

Used to make other chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2528-39-4
Molecular Formula C18H39O4P
Molecular Weight (g/mol) 350.48
MDL Number MFCD00015351
InChI Key SFENPMLASUEABX-UHFFFAOYSA-N
Synonym phosphoric acid, trihexyl ester, tri-n-hexyl phosphate, tri n-hexyl phosphate, acmc-20aje0, aronis24109
PubChem CID 75666
IUPAC Name trihexyl phosphate
SMILES CCCCCCOP(=O)(OCCCCCC)OCCCCCC

Specifications

Color Colorless
Linear Formula [CH3(CH2)5O]3PO
Quantity 5 g
Formula Weight 350.48
Percent Purity ≥90%
Physical Form Liquid
Chemical Name or Material Tri-n-hexyl phosphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.