missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-butyltin isocyanate, 97%, Thermo Scientific™

Product Code. 15481818
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15481818 25 g 25g
15461818 1 g 1g
15471818 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15481818 Supplier Thermo Scientific Alfa Aesar Supplier No. H55526.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 681-99-2
Molecular Formula C13H27NOSn
Molecular Weight (g/mol) 332.075
MDL Number MFCD08272321
InChI Key YGUNDMHXMXDGII-UHFFFAOYSA-N
Synonym tributyltin isocyanate, tributyl isocyanato stannane, stannane, isocyanato tributyl, tri-n-butyltin isocyanate, tributylstannyl isocyanate, tributyltin isothiocyanate, stannane, tributylisocyanato, tin, tributyl-, isothiocyanate, tri-n-butylisocyanatotin, acmc-20ajf1
PubChem CID 120323
IUPAC Name tributyl(isocyanato)stannane
SMILES CCCC[Sn](CCCC)(CCCC)N=C=O

Specifications

Color Colorless
Density 1.203 g/mL
Boiling Point 297°C
Flash Point 110°C (230°F)
Linear Formula [CH3(CH2)3]3SnNCO
Refractive Index 1.489
Quantity 25 g
UN Number UN2788
Formula Weight 332.05
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Tri-n-butyltin isocyanate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.