missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-butyltin chloride, 96%

Product Code. 11429632
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11429632 50 g 50g
11439632 250 g 250g
11419632 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11429632 Supplier Thermo Scientific Alfa Aesar Supplier No. A10746.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1461-22-9 | C12H27ClSn | 325.508 g/mol

Tri-n-butyltin chloride is used as an endocrine disruptor as well as an inhibitor for the V-ATPases. It is also used as a rodent-repellent for cable coatings. Further, it is used in hot end glass coating. It also employed as a corrosion remover.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tri-n-butyltin chloride is used as an endocrine disruptor as well as an inhibitor for the V-ATPases. It is also used as a rodent-repellent for cable coatings. Further, it is used in hot end glass coating. It also employed as a corrosion remover.

Solubility
Miscible with alcohol, heptane, benzene and toluene. Immiscible with water.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1461-22-9
Molecular Formula C12H27ClSn
Molecular Weight (g/mol) 325.508
MDL Number MFCD00000521
InChI Key GCTFWCDSFPMHHS-UHFFFAOYSA-M
Synonym tributyltin chloride, chlorotributyltin, tri-n-butyltin chloride, tributylchlorotin, stannane, tributylchloro, chlorotri-n-butyltin, tri-n-butylchlorotin, tributyl chloro stannane, chlorotributylstannane, tributylstannyl chloride
PubChem CID 15096
ChEBI CHEBI:79734
IUPAC Name tributyl(chloro)stannane
SMILES CCCC[Sn](CCCC)(CCCC)Cl

Specifications

Melting Point -19°C
Density 1.2
Boiling Point 171°C to 173°C (25 mmHg)
Flash Point 120°C (248°F)
Refractive Index 1.491
Quantity 50 g
UN Number UN2788
Beilstein 3535715
Solubility Information Miscible with alcohol,heptane,benzene and toluene. Immiscible with water.
Formula Weight 325.51
Percent Purity 96%
Chemical Name or Material Tri-n-butyltin chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.