missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-butylphosphine, 94%

Product Code. 15471057
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15471057 50 g 50g
15481057 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15471057 Supplier Thermo Scientific Alfa Aesar Supplier No. 014114.18
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 998-40-3 | C12H27P | 202.32 g/mol

Tri-n-butylphosphine is used as a catalyst modifier in the cobalt-catalyzed hydroformylation of alkenes for industrial use. It is also used as a strong reducing agent and polymerization cross-linking catalyst. It is a used as an intermediate for phase transfer catalysts. Used with platinum (II) or (IV) in an intermolecular hydroamination of inactivated alkenes. It is used as 1,4-addition catalyst; used with disulfides for the thioetherification of alcohols; acylation catalyst; used to prepare active esters.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • For cleavage of disulfide bonds, as a polymerization crosslinking catalyst, as an organic intermediate, and in fuels
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 998-40-3
Molecular Formula C12H27P
Molecular Weight (g/mol) 202.32
MDL Number MFCD00009462
InChI Key TUQOTMZNTHZOKS-UHFFFAOYSA-N
Synonym tributylphosphine, tri-n-butylphosphine, phosphine, tributyl, tris butyl phosphine, tributylfosfin, tributylfosfin czech, tributyl-phosphane, unii-0o52fjr7wn, tri-n-butyl phosphine, tributyl phosphine tbp
PubChem CID 13831
IUPAC Name tributylphosphane
SMILES CCCCP(CCCC)CCCC

Specifications

Melting Point -65°C
Density 0.820 g/mL
Boiling Point 240°C
Flash Point >93°C (199°F)
Odor Unpleasant
Refractive Index 1.462
Quantity 50 g
UN Number UN3254
Beilstein 1738261
Sensitivity Air and moisture sensitive
Solubility Information Insoluble in water; Miscible with ether, methanol, ethanol and benzene
Formula Weight 202.32
Percent Purity 94%
Physical Form Liquid
Chemical Name or Material Tri-n-butylphosphine
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tri-n-butylphosphine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Pyrophoric liquid Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H250-Catches fire spontaneously if exposed to air.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Tributylphosphine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.