missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-butylphenyltin, 97%, Thermo Scientific Chemicals

Product Code. 15471908
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is applied in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied in chemical research.

Solubility
Sparingly Soluble (9.9E-5 g/L) (25°C)

Notes
Store at room temperature. Avoid strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 960-16-7
Molecular Formula C18H32Sn
Molecular Weight (g/mol) 367.164
MDL Number MFCD00134394
InChI Key SYUVAXDZVWPKSI-UHFFFAOYSA-N
Synonym tributyl phenyl stannane, tributylphenyltin, phenyltributylstannane, stannane, tributylphenyl, phenyltributyltin, phenyltri-n-butyltin, phenyltributlytin, tributylphenyl tin, phenyl tnbutyl tin, phenyl tributyl tin
PubChem CID 607632
IUPAC Name tributyl(phenyl)stannane
SMILES CCCC[Sn](CCCC)(CCCC)C1=CC=CC=C1

Specifications

Color Colorless
Density 1.125 g/mL
Boiling Point 125°C to 128°C (0.14 mmHg)
Flash Point 113°C (235°F)
Linear Formula C6H5Sn[(CH2)3CH3]3
Refractive Index 1.516
Quantity 5 g
UN Number UN2788
Beilstein 3610571
Solubility Information Sparingly Soluble (9.9E-5 g/L) (25°C)
Formula Weight 367.14
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Tri-n-butylphenyltin
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'