missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-butylamine, 98%

Product Code. p-7022212
Click to view available options
Quantity:
10,000 mL
2500 mL
500 mL
Unit Size:
10000mL
2500mL
500mL
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 11452657

Brand: Thermo Scientific Alfa Aesar A14868.AP

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 102-82-9 | C12H27N | 185.355 g/mol

Tri-n-butylamine is an important intermediate in the production of phase transfer catalysts like tributylmethylammonium chloride and tributylbenzylammonium chloride. It is also used in pharmaceuticals, agrochemicals, surfactants, lubricant additives, vulcanization accelerators and dyes. It acts as a catalyst and as a solvent in organic syntheses and polymerization reactions. It serves as a strong base anion exchanger, acid acceptor, inhibitor in hydraulic fluids and an emulsifying agent. Further, it is used to prepare photographic chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tri-n-butylamine is an important intermediate in the production of phase transfer catalysts like tributylmethylammonium chloride and tributylbenzylammonium chloride. It is also used in pharmaceuticals, agrochemicals, surfactants, lubricant additives, vulcanization accelerators and dyes. It acts as a catalyst and as a solvent in organic syntheses and polymerization reactions. It serves as a strong base anion exchanger, acid acceptor, inhibitor in hydraulic fluids and an emulsifying agent. Further, it is used to prepare photographic chemicals.

Solubility
Miscible with acetone, ethanol, ethyl ether and most organic solvents. Immiscible with water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-82-9
Molecular Formula C12H27N
Molecular Weight (g/mol) 185.355
MDL Number MFCD00009431
InChI Key IMFACGCPASFAPR-UHFFFAOYSA-N
Synonym tributylamine, tri-n-butylamine, 1-butanamine, n,n-dibutyl, n,n-dibutyl-1-butanamine, tributilamina, tris-n-butylamine, amine, tributyl, tris n-butylamine, tributilamina romanian, unii-c3tzb2w0r7
PubChem CID 7622
ChEBI CHEBI:38905
IUPAC Name N,N-dibutylbutan-1-amine
SMILES CCCCN(CCCC)CCCC

Specifications

Melting Point <-70°C
Density 0.778
pH 10.6
Boiling Point 216°C
Flash Point 71°C (159°F)
Odor Amine-like
Linear Formula [CH3(CH2)3]3N
Refractive Index 1.429
Quantity 500 mL
UN Number UN2542
Beilstein 1698872
Merck Index 14,9618
Solubility Information Miscible with acetone,ethanol,ethyl ether and most organic solvents. Immiscible with water.
Formula Weight 185.36
Percent Purity 98%
Chemical Name or Material Tri-n-butylamine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.