missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-butyl phosphate, 98%, Thermo Scientific Chemicals

Product Code. 11433097
Change view
Click to view available options
Quantity:
250 mL
1000 mL
5000 mL
Unit Size:
1000mL
250mL
5000mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11433097 5000 mL 5000mL
11423097 250 mL 250mL
11413097 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11433097 Supplier Thermo Scientific Alfa Aesar Supplier No. A16084.0J
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Tri-n-butyl phosphate is used as a solvent for rare earth metals exaction and purification, and as a flame retardant component of aircraft hydraulic fluid. It is involved in the formation of stable hydrophobic complexes with some metals. It is also employed as a defoamer additive in cement casings for oil wells, as an anti-air entrainment additive for coatings and floor finishes and as a solvent in nuclear fuel processing. Further, it is used as a solvent in inks, synthetic resins, gums and adhesives. In addition, it is used as a plasticizer for cellulose esters like nitrocellulose and cellulose acetate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tri-n-butyl phosphate is used as a solvent for rare earth metals exaction and purification, and as a flame retardant component of aircraft hydraulic fluid. It is involved in the formation of stable hydrophobic complexes with some metals. It is also employed as a defoamer additive in cement casings for oil wells, as an anti-air entrainment additive for coatings and floor finishes and as a solvent in nuclear fuel processing. Further, it is used as a solvent in inks, synthetic resins, gums and adhesives. In addition, it is used as a plasticizer for cellulose esters like nitrocellulose and cellulose acetate.

Solubility
Miscible with diethyl ether, benzene, carbon disulfide and ethanol. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents, strong bases and alkali.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126-73-8
Molecular Formula C12H27O4P
Molecular Weight (g/mol) 266.32
MDL Number MFCD00009436
InChI Key STCOOQWBFONSKY-UHFFFAOYSA-N
Synonym tri-n-butyl phosphate, tributylphosphate, butyl phosphate, phosphoric acid tributyl ester, tributylphosphat, celluphos 4, disflamoll tb, tributilfosfato, tributylfosfaat, tributyle phosphate de
PubChem CID 31357
ChEBI CHEBI:35019
IUPAC Name tributyl phosphate
SMILES CCCCOP(=O)(OCCCC)OCCCC

Specifications

Melting Point -80°C
Density 0.977
Boiling Point 155°C to 156°C (10 mmHg)
Flash Point 152°C (305°F)
Assay Percent Range 98%
Odor Odorless
Refractive Index 1.424
Quantity 5000 mL
Beilstein 1710584
Merck Index 14,9618
Solubility Information Miscible with diethyl ether,benzene,carbon disulfide and ethanol. Slightly miscible with water.
Formula Weight 266.32
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Tri-n-butyl phosphate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tri-n-butyl phosphate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H351-Suspected of causing cancer.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Tributyl phosphate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.