missing translation for 'onlineSavingsMsg'
Learn More

Tri-n-butyl citrate, 99%

Product Code. 11493983
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11493983 50 g 50g
11403993 250 g 250g
11483983 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11493983 Supplier Thermo Scientific Alfa Aesar Supplier No. A10266.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 77-94-1 | C18H32O7 | 360.45 g/mol

Tri-n-butyl citrate is used in polishes, inks, nitrocellulose lacquers, as an anti-foam agent and as a plasticizer. It is also used as paint additives, coating additives and processing aids. In addition to this, it is added to the zein film to enhance its mechanical properties for industrial processing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tri-n-butyl citrate is used in polishes, inks, nitrocellulose lacquers, as an anti-foam agent and as a plasticizer. It is also used as paint additives, coating additives and processing aids. In addition to this, it is added to the zein film to enhance its mechanical properties for industrial processing.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-94-1
Molecular Formula C18H32O7
Molecular Weight (g/mol) 360.45
MDL Number MFCD00027217
InChI Key ZFOZVQLOBQUTQQ-UHFFFAOYSA-N
Synonym tributyl citrate, butyl citrate, tri-n-butyl citrate, n-butyl citrate, citroflex 4, citric acid, tributyl ester, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, tributyl ester, butyl citrate van, unii-827d5b1b6s, 2-hydroxy-1,2,3-propanetricarboxylic acid, tributyl ester
PubChem CID 6507
IUPAC Name tributyl 2-hydroxypropane-1,2,3-tricarboxylate
SMILES CCCCOC(=O)CC(O)(CC(=O)OCCCC)C(=O)OCCCC

Specifications

Melting Point -20°C
Density 1.04
Boiling Point 325°C
Flash Point 157°C (315°F)
Odor Weak
Refractive Index 1.444
Quantity 50 g
Beilstein 1806072
Merck Index 14,1564
Solubility Information Immiscible with water.
Formula Weight 360.45
Percent Purity 99%
Chemical Name or Material Tri-n-butyl citrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.