missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tri-iso-propyl-d21-phosphine Carbon Disulfide Complex, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Tri(isopropyl)phosphine-D21 Carbon Disulfide Complex |
| CAS | 1219798-41-0 |
| Molecular Formula | C9 D21 P . C S2 |
| Quantity | 0.25 g |
| Synonym | Phosphine, tris[1-(methyl-d3)ethyl-1,2,2,2-d4]-, compd. with carbon disulfide (1:1) (ACI), Tri-iso-propyl-d21-phosphine carbon disulfide complex, Tri(isopropyl)phosphine-D21 carbon disulfide complex, Tri(isopropyl-d7)phosphine carbon disulfide complex |
| InChI Formula | InChI=1 S/C9H21P.CS2/c1-7(2)10(8(3)4)9(5)6;2-1-3/h7-9 H,1-6H3;/i1D3,2D3,3D3,4D3,5D3,6D3,7 D,8 D,9 D; |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])(P(C([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H])C([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H].S=C=S |
| IUPAC Name | methanedithione;tris(1,1,1,2,3,3,3-heptadeuteriopropan-2-yl)phosphane |
| Molecular Weight (g/mol) | 257.507 |
| Formula Weight | 257.214 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction