missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ tri-GalNAc COOH

Product Code. 18736616
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18736616 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18736616 Supplier Tocris Bioscience™ Supplier No. 7401/1

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Asialoglycoprotein receptor ligand with PEG linker and carboxylic acid group for onward chemistry

Tri-GalNAc COOH is a functionalized asialoglycoprotein receptor ligand for lysosomal targeting chimera (LYTAC) research and development; incorporates an asialoglycoprotein receptor ligand with PEG linker and carboxylic acid group reactive handle ready for conjugation to a target protein ligand.
TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material 36-[[2-(Acetylamino)-2-deoxy-β-D-galactopyranosyl]oxy]-21,21-bis[[3-[[3-[[5-[[2-(acetylamino)-2-deoxy-β-D-galactopyranosyl]oxy]-1-oxopentyl]amino]propyl]amino]-3-oxopropoxy]methyl]-19,26,32-trioxo-4,7,10,13,16,23-hexaoxa-20,27,31-triazahexatriac
CAS 1953146-81-0
Quantity 1 mg
Target Degrader Building Blocks
Molecular Formula C75H134N10O35
Purity 0.9
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.