missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ tri-GalNAc-C5-amine
Description
tri-GalNAc-C5-amine is a functionalized asialoglycoprotein receptor (ASGPR) ligand for lysosomal targeting chimera (LYTAC) research and development; each molecule incorporates three ASGPR ligands with an alkylC5 linker and an amine group reactive handle ready for conjugation. Upon binding to the ASGPR, tri-GalNAc conjugates are efficiently internalized via ASGPR-mediated endocytosis. tri-GalNAc conjugation can be employed as a strategy to effectively deliver cargo such as RNA or Cas9 complexes in a cell-specific manner to hepatocytes. Can be used to generate LYTACs, or labeled with dye for tissue imaging.
Specifications
Specifications
| Chemical Name or Material | N-[2-[[1-[2-[2-[2-[2-[[2-(Acetylamino)-2-deoxy-β-D-galactopyranosyl]oxy]ethoxy]ethoxy]ethoxy]ethyl]-1H-1,2,3-triazol-4-yl]methoxy]-1,1-bis[[[1-[2-[2-[2-[2-[[2-(acetylamino)-2-deoxy-β-D-galactopyranosyl]oxy]ethoxy]ethoxy]ethoxy]ethyl]-1H-1,2,3-tr |
| CAS | 2098275-86-4 |
| Quantity | 1 mg |
| Product Line | Tocris |
| Target | ASGPR1 |
| Molecular Formula | C67H118N14O31 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?