missing translation for 'onlineSavingsMsg'
Learn More

Tri(ethylene glycol) di-p-toluenesulfonate, 98%

Product Code. 11490877
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11490877 10 g 10g
11400887 50 g 50g
11410887 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11490877 Supplier Thermo Scientific Alfa Aesar Supplier No. B23279.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19249-03-7 | C20H26O8S2 | 458.54 g/mol

Tri(ethylene glycol) di-p-toluenesulfonate is used in the synthesis of 1-[4-(1,4,7,10-tetraoxa-13-aza-cyclopentadec-13-yl)-phenyl] ethanone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tri(ethylene glycol) di-p-toluenesulfonate is used in the synthesis of 1-[4-(1,4,7,10-tetraoxa-13-aza-cyclopentadec-13-yl)-phenyl] ethanone.

Solubility
Soluble in ethanol.

Notes
Incompatible with strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19249-03-7
Molecular Formula C20H26O8S2
Molecular Weight (g/mol) 458.54
MDL Number MFCD00048096
InChI Key KCONMNWPRXAWKK-UHFFFAOYSA-N
Synonym tri ethylene glycol di-p-toluenesulfonate, triethylene glycol ditosylate, ethane-1,2-diylbis oxy bis ethane-2,1-diyl bis 4-methylbenzenesulfonate, triethylene glycol di p-toluenesulfonate, 1,2-bis 2-tosyloxyethoxy ethane, 2-2-2-4-methylbenzenesulfonyl oxy ethoxy ethoxy ethyl 4-methylbenzene-1-sulfonate, ethane-1,2-diylbis oxyethane-2,1-diyl bis 4-methylbenzenesulfonate, 2-2-2-4-methylphenyl sulfonyloxy ethoxy ethoxy ethyl 4-methylbenzenesulfon ate, 2-2-2-4-methylphenyl sulfonyloxyethoxy ethoxy ethyl 4-methylbenzenesulfonate, 2-2-2-4-methylbenzenesulfonyl oxy ethoxy ethoxy ethyl 4-methylbenzenesulfonate
PubChem CID 316652
IUPAC Name 2-[2-[2-(4-methylphenyl)sulfonyloxyethoxy]ethoxy]ethyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OCCOCCOCCOS(=O)(=O)C2=CC=C(C=C2)C

Specifications

Melting Point 80°C to 82°C
Quantity 10 g
Solubility Information Soluble in ethanol.
Formula Weight 458.55
Percent Purity 98%
Chemical Name or Material Tri(ethylene glycol) di-p-toluenesulfonate
compliance-icons
Product Identifier
  • Tri(ethylene glycol) di-p-toluenesulfonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Triethylene glycol di(p-toluenesulfonate)

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.