Learn More
BOLA Tri-Clamp GL-Adaptor
Product Code. p-200109944
Click to view available options
Diameter (Metric) Inner:
16 mm
32 mm
Unit Size:
Each
Product Code. 15513293
Brand: BOLA D90124
Only null left
Add to basket
Add to basket
Tri-Clamp GL-Adaptor Fitting for transition of Tri-Clamp as per DIN 32 676
  • Fitting made of PTFE for transition of Tri-Clamp as per DIN 32 676 to a GL-threaded neck.
  • Universal chemical resistance, the flowing product is only exposed to PTFE.
Specifications
Certifications/Compliance FDA
Autoclavable Autoclavable
Diameter (Metric) Inner 16 mm
Thread Size GL 18
Color White