missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BOLA Tri-Clamp GL-Adaptor
Click to view available options
Diameter (Metric) Inner:
16 mm
32 mm
Unit Size:
Each
Description
- Fitting made of PTFE for transition of Tri-Clamp as per DIN 32 676 to a GL-threaded neck.
- Universal chemical resistance, the flowing product is only exposed to PTFE.
Specifications
Specifications
| Certifications/Compliance | FDA |
| Autoclavable | Autoclavable |
| Diameter (Metric) Inner | 16 mm |
| Thread Size | GL 18 |
| Color | White |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction