missing translation for 'onlineSavingsMsg'
Learn More

tri(2-methylphenyl)phosphine, Thermo Scientific™

Product Code. 10422203
Click to view available options
Quantity:
10 g
50 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 6163-58-2
Molecular Formula C21H21P
Molecular Weight (g/mol) 304.373
InChI Key COIOYMYWGDAQPM-UHFFFAOYSA-N
Synonym tri-o-tolylphosphine, tri o-tolyl phosphine, tris 2-methylphenyl phosphine, tris 2-methylphenyl phosphane, phosphine, tris 2-methylphenyl, tris o-tolyl phosphine, tri-ortho-toylphosphine, tri-ortho-tolylphosphine, phosphine, tri-o-tolyl, tri 2-methylphenyl phosphine
PubChem CID 80271
IUPAC Name tris(2-methylphenyl)phosphane
SMILES CC1=CC=CC=C1P(C2=CC=CC=C2C)C3=CC=CC=C3C

Specifications

Assay Percent Range 97%
Linear Formula (CH3C6H4)3P
Quantity 10 g
Formula Weight 304.37
Percent Purity 97%
Chemical Name or Material Tri(2-methylphenyl)phosphine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'