missing translation for 'onlineSavingsMsg'
Learn More

Tri(1-naphthyl)phosphine, 97%

Product Code. 11394348
Change view
Click to view available options
Quantity:
0.5 g
2 g
10 g
50 g
Unit Size:
0.5g
10g
2g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11394348 10 g 10g
11314358 0.5 g 0.5g
11304358 2 g 2g
11384348 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11394348 Supplier Thermo Scientific Alfa Aesar Supplier No. 042304.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3411-48-1 | C30H21P | 412.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3411-48-1
Molecular Formula C30H21P
Molecular Weight (g/mol) 412.47
MDL Number MFCD00046400
InChI Key DMEUUKUNSVFYAA-UHFFFAOYSA-N
Synonym tri 1-naphthyl phosphine, tri-1-naphthylphosphine, tri naphthalen-1-yl phosphine, tris 1-naphthyl phosphine, tris naphthalen-1-yl phosphane, trinaphthyl phosphine, tri-1-naphtylphosphine, tri-1-napthylphosphine, acmc-1cp9e, tri naphthalen-1-yl phosphane
PubChem CID 271443
IUPAC Name trinaphthalen-1-ylphosphane
SMILES C1=CC2=CC=CC(P(C3=CC=CC4=CC=CC=C34)C3=C4C=CC=CC4=CC=C3)=C2C=C1

Specifications

Melting Point 265°C to 268°C
Quantity 10 g
Formula Weight 412.47
Physical Form Powder
Chemical Name or Material Tri (1-naphthyl)phosphine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.