missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRHR Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA141469
This item is not returnable.
View return policy
Description
TRHR Polyclonal Antibody for Western Blot, IHC (P)
TRH-R is a 398 amino acid integral membrane protein with six transmembrane domains and belongs to G-protein coupled receptor 1 family. It acts as a receptor for the signaling hormone TRH and activates the inositol phospholipid-calcium-protein kinase C transduction pathway upon binding with TRH. Reports suggest that TRH-R is a neurotransmitter and neuro regulator that controls dimerization of the TRH receptor and potentiates hormone-induced receptor trafficking. It is known to widely express in the thyrotroph cells of the anterior pituitary.
Specifications
| TRHR | |
| Polyclonal | |
| Unconjugated | |
| TRHR | |
| MGC141920; Thyroliberin receptor; thyrotropin releasing hormone receptor; thyrotropin-releasing hormone receptor; TRH receptor; TRH1 Receptor; Trhr; TRH-R; TRH-R1 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 482007, 7201 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.2% gelatin and 0.05% sodium azide | |
| P34981 | |
| TRHR | |
| A portion of amino acids 350-400 of human TRH-R protein. | |
| 100 μg | |
| Primary | |
| Human, Canine, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction