missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRHDE Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA547429
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
This gene encodes a member of the peptidase M1 family. TRHDE is an extracellular peptidase that specifically cleaves and inactivates the neuropeptide thyrotropin-releasing hormone.
Specifications
| TRHDE | |
| Polyclonal | |
| Unconjugated | |
| TRHDE | |
| 9330155P21Rik; PAP-II; PGPEP2; pyroglutamyl aminopeptidase II; pyroglutamyl-peptidase II; Thyroliberinase; thyrotropin releasing hormone degrading enzyme; thyrotropin-releasing hormone degrading ectoenzyme; thyrotropin-releasing hormone degrading enzyme; thyrotropin-releasing hormone-degrading ectoenzyme; Trhde; TRH-DE; TRH-degrading ectoenzyme; TRH-degrading enzyme; TRH-specific aminopeptidase; UNQ2507/PRO5995 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 237553, 366894 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q10836, Q8K093 | |
| TRHDE | |
| Mouse myeloma cell line NS0-derived recombinant mouse TRH-degrading Ectoenzyme/TRHDE Arg64-His1025. | |
| 100 μg | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction