missing translation for 'onlineSavingsMsg'
Learn More

TRH-degrading Ectoenzyme/TRHDE Antibody [DyLight 680], Novus Biologicals™

Product Code. 30661964 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30661964 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30661964 Supplier Novus Biologicals Supplier No. AF2985Y

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

TRH-degrading Ectoenzyme/TRHDE Polyclonal antibody specifically detects TRH-degrading Ectoenzyme/TRHDE in Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRH-degrading Ectoenzyme/TRHDE
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias EC 3.4.19.6, FLJ22381, PAP-IIthyroliberinase, PGPEP2, Pyroglutamyl-peptidase II, Thyroliberinase, thyrotropin-releasing hormone degrading enzyme, thyrotropin-releasing hormone-degrading ectoenzyme, TRH-degrading ectoenzyme, TRH-DEthyrotropin-releasing hormone degrading ectoenzyme, TRH-specific aminopeptidase
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse TRH-degrading Ectoenzyme/TRHDE, Arg64-His1025, Accession # Q8K093
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 29953
Target Species Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.