missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TRF4-1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA129801
This item is not returnable.
View return policy
Description
Detects a band ~92 kDa.
TRF4-1 is a nuclear protein whose expression is cell cycle-regulated at a post-transcriptional level. Two genes, TRF4-1 and TRF4-2, belonging to this family have been identified. TRF4 is also required to maintain a different aspect of higher order chromosome structure, sister chromatid cohesion.
Specifications
| TRF4-1 | |
| Polyclonal | |
| Unconjugated | |
| TENT4A | |
| DNA polymerase kappa; DNA polymerase sigma; LAK1; LAK-1; non-canonical poly(A) RNA polymerase PAPD7; PAP associated domain containing 7; PAP-associated domain-containing protein 7; Papd7; POLK; POLS; polymerase (DNA directed) sigma; polymerase (DNA-directed) sigma; Tent4a; Terminal guanylyltransferase; Terminal nucleotidyltransferase 4A; Terminal uridylyltransferase 5; topoisomerase-related function protein 4-1; TRAMP-like complex polyadenylate polymerase; TRF4; TRF41; TRF4-1; TUTase 5; TUTASE5 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 11044 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q5XG87 | |
| TENT4A | |
| Synthetic peptide containing amino acids 447-462 (GTASLKAVHHMSSPAI) of human TRF4-1. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction