missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TRF1 Monoclonal Antibody (4E4)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA531596

Product Code. 16305034

  • £444.00 / 1mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Keep as concentrated solution.

This gene encodes a telomere specific protein which is a component of the telomere nucleoprotein complex. This protein is present at telomeres throughout the cell cycle and functions as an inhibitor of telomerase, acting in cis to limit the elongation of individual chromosome ends. The protein structure contains a C-terminal Myb motif, a dimerization domain near its N-terminus and an acidic N-terminus. Two transcripts of this gene are alternatively spliced products.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TRF1
Monoclonal
Conc. Not Determined
Tissue culture supernatant with no preservative
P54274
TERF1
1 mL
Primary
Human
Antibody
IgG1
ChIP Assay, Immunoprecipitation, Western Blot, Immunocytochemistry
4E4
Unconjugated
TERF1
FLJ41416; hTRF1-AS; NIMA-interacting protein 2; PIN2; Telomeric protein Pin2/TRF1; telomeric repeat binding factor (NIMA-interacting) 1; telomeric repeat binding factor 1; telomeric repeat-binding factor 1; TERF1; TRBF1; TRF; TRF1; TTAGGG repeat-binding factor 1; t-TRF1
Mouse
RUO
7013
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.