missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRF-2 Antibody (4A794.15), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-56506PE
This item is not returnable.
View return policy
Description
TRF-2 Monoclonal specifically detects TRF-2 in Human, Mouse, Rat, Deer, Mammal samples. It is validated for Flow Cytometry.
Specifications
| TRF-2 | |
| Monoclonal | |
| PE | |
| Q15554 | |
| TERF2 | |
| This TRF-2 Antibody (4A794.15) [PE] was developed against Baculovirus expressed whole length TRF2 protein, used for immunizing mice and splenocytes used to generate the hybridoma clone (NP_005643). | |
| Protein G purified | |
| RUO | |
| Primary | |
| The TRF2 antibody recognizes full-length TRF2 as well as TRF2 forms lacking both the N-terminal basic domain (B) and the telobox Myb-like C-terminal DNA-binding domain (M). TRF2 forms missing the B and M domains are often referred to as mutant TRF2. Although the exact epitope recognized by the TRF2 antibody has not been mapped, the scientific literature indicates it is in the D or L domain, but not in the B or M domain. | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Flow Cytometry | |
| 4A794.15 | |
| Flow Cytometry | |
| Telomeric DNA-binding protein, telomeric repeat binding factor 2, Telomeric repeat binding protein 2, telomeric repeat-binding factor 2, TERF 2, TERF2, TRBF 2, TRBF2, TRF 2, TRF2, TTAGGG repeat binding factor 2, TTAGGG repeat-binding factor 2 | |
| Mouse | |
| 59.6 kDa | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Transcription Factors and Regulators | |
| 7014 | |
| Human, Mouse, Rat, Deer, Mammalia | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction