missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRF-1 Antibody (57-6), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-68281PE
This item is not returnable.
View return policy
Description
TRF-1 Monoclonal specifically detects TRF-1 in Human, Mouse, Rat, Monkey samples. It is validated for Flow Cytometry.
Specifications
| TRF-1 | |
| Monoclonal | |
| PE | |
| PBS with 0.05% Sodium Azide | |
| TERF1 | |
| Recombinant human TRF1 [UniProt# P54274] | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Purified |
| Flow Cytometry | |
| 57-6 | |
| Flow Cytometry | |
| NIMA-interacting protein 2, PIN2hTRF1-AS, Telomeric protein Pin2/TRF1, telomeric repeat binding factor (NIMA-interacting) 1, telomeric repeat binding factor 1, telomeric repeat binding protein 1, telomeric repeat-binding factor 1, TERF1, TRBF1, TRF1FLJ41416, TRFt-TRF1, TTAGGG repeat-binding factor 1 | |
| Mouse | |
| Protein G purified | |
| Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Core ESC Like Genes, Stem Cell Markers | |
| 7013 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction