missing translation for 'onlineSavingsMsg'
Learn More

Treprostinil, MedChemExpress

Product Code. 16450016
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
2mg
5mg
This item is not returnable. View return policy

Product Code. 16450016

Brand: Medchem Express HY100441

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Treprostinil (UT-15) is a potent DP1 and EP2 agonist with EC50 values of 0.6±0.1 and 6.2±1.2 nM, respectively.

Treprostinil (UT-15) is a potent DP1 and EP2 agonist with EC50 values of 0.6±0.1 and 6.2±1.2 nM, respectively.

Specifications

Chemical Name or Material Treprostinil
CAS 81846-19-7
Color White
Purity Grade Notes Research
Molecular Formula C23H34O5
Quantity 2 mg
Synonym UT-15
Solubility Information DMSO : ≥ 125 mg/mL (320.09 mM)
SMILES OC(COC1=CC=CC2=C1C[C@](C[C@@H](O)[C@@H]3CC[C@@H](O)CCCCC)([H])[C@]3([H])C2)=O
Molecular Weight (g/mol) 390.51
Formula Weight 390.51
Percent Purity 98.71%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.