missing translation for 'onlineSavingsMsg'
Learn More

Treprostinil, MedChemExpress

Product Code. 16473018
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
2mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16473018 10 mg 10mg
16455757 5 mg 5mg
16450016 2 mg 2mg
16422586 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16473018 Supplier Medchem Express Supplier No. HY100441
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Treprostinil (UT-15) is a potent DP1 and EP2 agonist with EC50 values of 0.6±0.1 and 6.2±1.2 nM, respectively.

Treprostinil (UT-15) is a potent DP1 and EP2 agonist with EC50 values of 0.6±0.1 and 6.2±1.2 nM, respectively.

Specifications

Chemical Name or Material Treprostinil
CAS 81846-19-7
Color White
Purity Grade Notes Research
Molecular Formula C23H34O5
Quantity 10 mg
Synonym UT-15
Solubility Information DMSO : ≥ 125 mg/mL (320.09 mM)
SMILES OC(COC1=CC=CC2=C1C[C@](C[C@@H](O)[C@@H]3CC[C@@H](O)CCCCC)([H])[C@]3([H])C2)=O
Molecular Weight (g/mol) 390.51
Formula Weight 390.51
Percent Purity 98.71%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.