missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Treponema pallidum Monoclonal Antibody (2124)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA533297
This item is not returnable.
View return policy
Description
Does not cross react with Borrelia burgdorferi. Does not stain the entire bacterial cell.
This antibody reacts with ultrasonicated lysates of T. pallidum and T. reiter.
Specifications
| Treponema pallidum | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Mouse | |
| 1 mL | |
| Primary | |
| 4°C | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 2124 | |
| Unconjugated | |
| Syphillis; t pallidum; T. pallidum | |
| Protein A | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction