missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Treponema pallidum Haemagglutination Test Kit (TPHA)

Product Code. 13245639
Change view
Click to view available options
Quantity:
200 Tests
Unit Size:
200 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13245639 200 Tests 200 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13245639 Supplier Thermo Scientific™ Supplier No. DR0530M

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Serologically diagnose Syphilis with this haemagglutination test for Treponema pallidum.

Serologically diagnose Syphilis with Thermo Scientific™ Oxoid™ Treponema pallidum Haemagglutination Test (TPHA).  The haemagglutination test utilizes specific antigens for the detection of antibodies to Treponema pallidum in the diagnosis of Syphilis.

Syphilis is a sexually transmitted disease. The causative organism is Treponema pallidum, a spirochete which cannot be grown on culture media or in tissue culture. Diagnosis of infection is normally by the detection of antibody specific for Treponema pallidum in the patient’s blood or CSF.

The TPHA test kit is a sensitive passive haemagglutination test specifically designed for the detection of antibodies to Treponema pallidum.

  • Rapid: within little over 30 minutes of receiving the test sample or culture
  • Simple: ready-to-use reagents
  • Easy-to-read results
  • Excellent sensitivity: high quality specific antigens used in the test provide excellent sensitivity.

Tanned fowl erythrocytes are coated with specific antigen and suspended in a diluent 1,2,3. When diluted positive samples are mixed with the test suspension, antibody to the sensitising antigen causes agglutination of the cells. The cells form a characteristic pattern in the bottom of a microtitration plate well. In the absence of reacting antibody, the cells form a compact button in the well. Uncoated tanned fowl erythrocytes are used as the control cells.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Control Sets Positive
Description Treponema pallidum Haemagglutination Test Kit (TPHA)
Detectable Analytes T. pallidum Antibodies
Type Treponema pallidum Haemagglutination Test Kit (TPHA)
Includes Test cell suspension, control cell suspension, diluent buffer, positive control serum, negative control serum
Quantity 200 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.