Learn More
Emma Safety Footwear Trento Safety Shoes
Product Code. 13294434
Change view
Click to view available options
Size:
39
40
47
Unit Size:
1 pair
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
13294434 39 1 pair
13204444 40 1 pair
13274444 47 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 13294434 Supplier Emma Safety Footwear Supplier No. MM10509039
  • ONLINE EXCLUSIVE PRICE
    £158.55
    £106.24 / 1 pairs
    Save £52.31
    32% Off
Estimated Shipment: 10-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The formally styled business shoe that protects the feet from above and below with water-resistant leather uppers, zero penetration in-sole and a steel safety toe cap. Specifically designed for standing, walking and the occapional run.
  • S3-Safety shoe with sole plate for wet work
  • Full grain leather construction
  • Nitrile Rubber Heat Resistant Outsole
  • ESD Certified
  • Chrome-free lining
  • Sanitized
Specifications
Certifications/Compliance EN ISO 20345:2011, EN 12568:2010
Closure Type Laces
Color Black
Design Trento
Liner Material Hydro-Tec™ Sanitized Silver & Wellness leather
Material Leather
Size 39
Toe Material Steel