Learn More
uvex Trend BOA™ Black/Gray/Red PU/PU 69082 Low Shoe - Width 11
Product Code. 17273301 Shop All Uvex Products
Click to view available options
Size:
38
39
40
41
42
43
44
45
46
47
48
49
50
51
52
Unit Size:
1 pair
Product Code. 17273301
Brand: uvex 6908245
Only null left
Add to basket
Add to basket
Uvex 2 Trend safety footwear for medium-risk applications (S1-S3); heel cap offers increased stability without limiting movement
Uvex 2 Trend Perforated Shoe with BOA™ Fit System 6908.2 S1 P SRC
  • Sporty, S1 P perforated safety shoe with wide fit
  • Breathable perforated microvelour
  • Made from synthetic materials, suitable for people allergic to chrome
  • All sole materials free of silicones, plasticizers and other paint wetting impairment substances
  • BOA™ Fit System Dial for custom comfort that's consistent all day
  • Meets ESD specification with volume resistance under 35 megohms
  • Reliable protection for wearers
  • Steel toe cap and stabilizing Uvex anti-twist heel cap to prevent ankle twisting
  • Ergonomically designed outsole made from dual-density polyurethane with excellent slip resistance
  • Reflective details for enhanced visibility
  • Uvex Climazone technology lining, upper and removable comfortable climatic insole with a moisture transport system
    • Helps maintain foot temperature for optimum wearer comfort
  • Soft padding on collar and tongue
  • Removable anti-static comfortable insole with moisture transport system and additional shock absorption around heel and forefoot
  • Article No. 6908.2
  • Standard EN ISO 20345:2011 S1 P SRC
  • Sizes: 38 to 52
TRUSTED_SUSTAINABILITY
Specifications
Color Black, Gray, Red
Material Microvelour
Size 45
Sole Material Dual-density polyurethane (PU/PU)
Toe Material Steel
Disposable Reusable
Certifications/Compliance (Europe) EN ISO 20345:2011
Gender Unisex
Fit Standard Width
Model Uvex 2
Latex Free Yes
Waterproof No
EN ISO 20345 S1
Slip Resistance Rating SRC
Product Type ESD Safety Shoe
Show More Show Less
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.