Learn More
uvex Trend BOA™ Black/Blue PU/PU 69382 Low Shoe - Width 11
Product Code. 17253911 Shop All Uvex Products
Change view
Click to view available options
Size:
38
39
40
41
42
43
44
45
46
47
48
49
50
51
52
Unit Size:
1 pair
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17253911 45 1 pair
17283901 38 1 pair
17293901 39 1 pair
17203911 40 1 pair
17213911 41 1 pair
17223911 42 1 pair
17233911 43 1 pair
17243911 44 1 pair
17263911 46 1 pair
17273911 47 1 pair
17283911 48 1 pair
17293911 49 1 pair
17203921 50 1 pair
17213921 51 1 pair
17223921 52 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Product Code. 17253911 Supplier uvex Supplier No. 6938245
  • ONLINE EXCLUSIVE PRICE
    £112.00
    £98.33 / 1 pairs
    Save £13.67
    12% Off
View all promotions
Estimated Shipment: 17-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Uvex 2 Trend safety footwear for medium-risk applications (S1-S3); heel cap offers increased stability without limiting movement
Uvex 2 Trend Perforated Shoe with BOA™ Fit System 69382 S1 P SRC
  • Sporty S1 P perforated safety shoe with wide fit
  • Breathable, perforated microvelour
  • Made from synthetic materials, suitable for people allergic to chrome
  • All sole materials free of silicones, plasticizers and other paint wetting impairment substances
  • BOA™ Fit System Dial for custom comfort that's consistent all day
  • Meets ESD specification with volume resistance under 35 megohms
  • Reliable protection for wearers
  • Steel toecap and stabilizing Uvex anti-twist heel cap to prevent ankle twisting
  • Ergonomically designed outsole made from dual-density polyurethane with excellent slip resistance
  • Reflective details for enhanced visibility
  • Uvex Climazone technology lining, upper and removable comfortable climatic insole
    • Moisture transport system helps maintain foot temperature
  • Soft padding collar and dust tongue
  • Removable and anti-static comfortable insole with a moisture transport system and additional shock absorption around heel and forefoot
  • Article No. 69382
  • Standard EN ISO 20345:2011 S1 P SRC
  • Sizes: 38 to 52
TRUSTED_SUSTAINABILITY
Specifications
Color Black, Blue
Material Microvelour
Size 45
Sole Material Dual-density polyurethane (PU/PU)
Toe Material Steel
Disposable Reusable
Certifications/Compliance (Europe) EN ISO 20345:2011
Gender Unisex
Fit Standard Width
Model Uvex 2
Latex Free Yes
Waterproof No
EN ISO 20345 S1
Slip Resistance Rating SRC
Product Type ESD Safety Shoe