missing translation for 'onlineSavingsMsg'
Learn More

Trenbolone, Mikromol™

Product Code. 18090870
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18090870 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18090870 Supplier Mikromol Supplier No. MM1377.00
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Trenbolone
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trenbolone
CAS 10161-33-8
Analyte or Component Names Trenbolone
Shipping Condition Ice pack (-20°C)
Molecular Formula C18 H22 O2
Certifications/Compliance ISO 17034
Synonym (17beta)-17-Hydroxy-estra-4,9,11-trien-3-one, 17beta-Hydroxy-estra-4,9,11-trien-3-one, (+)-Trenbolone, 17beta-Hydroxyestra-4,9,11-trien-3-one, 17beta-Trenbolone, 9,10,11,12-Dehydro-19-nortestosterone, RU 2341, Trenbolone, Trienbolone, beta-Trenbolone, Trenbolone Acetate Related Compound B
InChI Formula InChI=1S/C18H22O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h8-10,15-17,20H,2-7H2,1H3/t15-,16+,17+,18+/m1/s1
SMILES C[C@]12C=CC3=C4CCC(=O)C=C4CC[C@H]3[C@@H]1CC[C@@H]2O
IUPAC Name (8S,13S,14S,17S)-17-hydroxy-13-methyl-2,6,7,8,14,15,16,17-octahydro-1H-cyclopenta[a]phenanthren-3-one
Molecular Weight (g/mol) 270.37
Quantity 100 mg
Formula Weight 270.162 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.