missing translation for 'onlineSavingsMsg'
Learn More

Trenbolone Acetate, LoGiCal

Product Code. 30459298
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30459298 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30459298 Supplier LoGiCal Supplier No. LGCFOR1459.00

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Spezifikation

Chemical Name or Material Trenbolone Acetate
CAS 10161-34-9
Analyte or Component Names Trenbolone Acetate
Shipping Condition Ice pack (-20°C)
Molecular Formula C20 H24 O3
Certifications/Compliance ISO 9001
Synonym Estra-4,9,11-trien-3-one, 17β-hydroxy-, acetate (7CI,8CI), (17β)-17-(Acetyloxy)estra-4,9,11-trien-3-one, 17β-Acetoxy-Δ4,9,11-estratrien-3-one, 17β-Acetoxyestra-4,9,11-trien-3-one, 17β-Hydroxyestra-4,9,11-trien-3-one 17-acetate, 3-Oxo-17β-acetoxyestra-4,9,11-triene, Component T-H, Component T-S, Finaplix, Finaplix H, Finaplix-S, RU 1697, Trenbolone acetate, Trienbolone acetate
InChI Formula InChI=1S/C20H24O3/c1-12(21)23-19-8-7-18-17-5-3-13-11-14(22)4-6-15(13)16(17)9-10-20(18,19)2/h9-11,17-19H,3-8H2,1-2H3/t17-,18+,19+,20+/m1/s1
Solution Type Neat
SMILES CC(=O)O[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CCC4=C3C=C[C@]12C
IUPAC Name [(8S,13S,14S,17S)-13-methyl-3-oxo-2,6,7,8,14,15,16,17-octahydro-1H-cyclopenta[a]phenanthren-17-yl] acetate
Industry Type Clinical Testing
Molecular Weight (g/mol) 312.4
Quantity 10 mg
Formula Weight 312.1725 g/mol
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.