missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TREML4/TLT-4 Antibody (817914) [PerCP], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB5500C
This item is not returnable.
View return policy
Description
TREML4/TLT-4 Monoclonal antibody specifically detects TREML4/TLT-4 in Mouse samples. It is validated for Flow Cytometry, CyTOF
Specifications
| TREML4/TLT-4 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 285852 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry, CyTOF | |
| 817914 | |
| Flow Cytometry, CyTOF-ready | |
| TLT4 | |
| Mouse myeloma cell line NS0-derived recombinant mouse TREML4/TLT-4, Gly25-Pro195, Accession # Q3LRV9 | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction