missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TREML4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564198
This item is not returnable.
View return policy
Description
Immunogen sequence: TCCFHLCCCC SWPQGAVPEE LHKHPGQTLL LQCQYSPKRG PYQPKSWCQQ TSPSRCTL Highest antigen sequence identity to the following orthologs: Mouse - 47%, Rat - 47%.
Positively regulates Toll-like receptor TLR7 signaling in macrophages.
Specifications
| TREML4 | |
| Polyclonal | |
| Unconjugated | |
| TREML4 | |
| 5031403H21Rik; BB137214; IDCP1; IG-like domain containing protein IDCP1c; TLT4; TLT-4; TREM; TREM like transcript 4; Treml3; Treml4; trem-like transcript 4 protein; Triggering receptor expressed on myeloid cells; triggering receptor expressed on myeloid cells like 4; triggering receptor expressed on myeloid cells-like 3; triggering receptor expressed on myeloid cells-like 3b; triggering receptor expressed on myeloid cells-like 4; triggering receptor expressed on myeloid cells-like protein 3; Triggering receptor expressed on myeloid cells-like protein 4; UNQ9425/PRO34675 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 285852 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6UXN2 | |
| TREML4 | |
| Recombinant protein corresponding to Human TREML4. Recombinant protein control fragment (Product #RP-103583). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction