Learn More
TREML1 (TLT-1) Rat anti-Human, Brilliant Violet 480, Clone: 268420, BD OptiBuild™
Rat Monoclonal Antibody
Brand: BD Biosciences 748122
This item is not returnable.
View return policy
Description
TREML1 (TLT-1) Monoclonal antibody specifically detects TREML1 (TLT-1) in Human samples. It is validated for Flow Cytometry.
Specifications
| TREML1 (TLT-1) | |
| Monoclonal | |
| 0.2 mg/mL | |
| Rat | |
| RUO | |
| TREML1 (TLT-1) | |
| Refrigerate (2 to 8°C) |
| Flow Cytometry | |
| 268420 | |
| Brilliant Violet 480 | |
| 50 μg | |
| Primary | |
| Human | |
| IgG2b κ |