missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ TREML1/TLT-1 Antibody Pair
Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
Specifications
Specifications
| Assay Range | 28.13 to 1800 pg/mL (example only, lot dependent) |
| Assay Sensitivity | 28.13 pg/mL (example only; lot dependent) |
| Conjugate | Unconjugated |
| Detection Method | Sandwich ELISA |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Product Type | Sandwich ELISA |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 340205 |
| Gene Symbol | TREML1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction