missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TREML1 Recombinant Rabbit Monoclonal Antibody (037), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£218.00 - £436.00
Specifications
| Antigen | TREML1 |
|---|---|
| Clone | 037 |
| Concentration | 1 mg/mL |
| Applications | ELISA |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17739883
|
Invitrogen™
MA543535 |
50 μL |
£218.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17719703
![]() |
Invitrogen™
MA543151 |
100 μL |
£436.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Specificity: Human TLT-1/TREML1
TREML1 is located in a gene cluster on chromosome 6 with the single Ig variable domain activating receptors TREM1 and TREM2, but it has distinct structural and functional properties. TREML1 enhances calcium signaling in an SHP2 -dependent manner.Specifications
| TREML1 | |
| 1 mg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative | |
| 5430401J17Rik; dJ238O23.3; GLTL1825; MGC119173; OTTHUMP00000017858; PRO3438; RGD1565175; Tlt1; TLT-1; TREM; TREML1; trem-like transcript 1 protein; Triggering receptor expressed on myeloid cells; triggering receptor expressed on myeloid cells like 1; triggering receptor expressed on myeloid cells-like 1; triggering receptor expressed on myeloid cells-like 1c; triggering receptor expressed on myeloid cells-like 1d; triggering receptor expressed on myeloid cells-like 1e; triggering receptor expressed on myeloid cells-like protein 1; UNQ1825/PRO3438 | |
| TREML1 | |
| IgG | |
| Protein A | |
| Antibody |
| 037 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q86YW5 | |
| 340205 | |
| Recombinant Human TLT-1/TREML1 Protein. | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| TREML1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title