missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TREM2b Antibody (RM0139-5J46), CoraFluor™ 1, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-22515CL1
This item is not returnable.
View return policy
Description
TREM2b Monoclonal antibody specifically detects TREM2b in Mouse samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| TREM2b | |
| Monoclonal | |
| CoraFluor 1 | |
| Trem2a, Trem2b, Trem2c, TREM-2triggering receptor expressed on myeloid cells 2a, Triggering receptor expressed on monocytes 2, triggering receptor expressed on myeloid cells 2 | |
| Mouse recombinant TREM2b (extracellular domain) | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| RM0139-5J46 | |
| PBS | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| 54209 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction