missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TREM2 Monoclonal Antibody (78.18), FITC

Rat Monoclonal Antibody
Brand: Invitrogen™ MA528223
This item is not returnable.
View return policy
Description
Rat anti Mouse TREM-2 antibody, clone 78.18 recognizes triggering receptor expressed on myeloid cells 2 (TREM-2), an activating receptor of the Ig superfamily.
TREM2 is a membrane protein that forms a receptor signaling complex with TYROBP. The encoded protein may be involved in chronic inflammation by triggering the production of constitutive inflammatory cytokines. Defects in this gene are a cause of polycystic lipomembranous osteodysplasia with sclerosing leukoencephalopathy (PLOSL).
Specifications
| TREM2 | |
| Monoclonal | |
| 0.1 mg/mL | |
| PBS with 1% BSA and 0.09% sodium azide | |
| Q99NH8 | |
| Trem2 | |
| Mouse TREM-2 Fc fusion protein. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG1 |
| Flow Cytometry | |
| 78.18 | |
| FITC | |
| Trem2 | |
| OTTHUMP00000039771; soluble TREM 2; soluble TREM2; sTREM 2; sTREM2; TREM; Trem2; TREM-2; Trem2a; Trem2b; Trem2c; Triggering receptor expressed on monocytes 2; Triggering receptor expressed on myeloid cells; triggering receptor expressed on myeloid cells 2; triggering receptor expressed on myeloid cells 2a; triggering receptor expressed on myeloid cells 2b; triggering receptor expressed on myeloid cells 2c | |
| Rat | |
| Protein G | |
| RUO | |
| 83433 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction