missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TREM2 Polyclonal antibody specifically detects TREM2 in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | TREM2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS, 1% BSA, 20% Glycerol (pH7) |
| Gene Alias | Trem2a, Trem2b, Trem2c, TREM-2triggering receptor expressed on myeloid cells 2a, Triggering receptor expressed on monocytes 2, triggering receptor expressed on myeloid cells 2 |
| Host Species | Rabbit |
| Immunogen | Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human TREM2 (R47H Mutant). The exact sequence is proprietary. |
| Purification Method | Antigen Affinity-purified |
| Quantity | 100 μg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?